Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VRNKTVEEV YVEMLKPRDG VRLKVQYRPE EFTKAKGLPG DSFYIRALYD RLADVEQELS FKKDDILYVD DTLPQGTFGS WMAWQLDENA QKIQRGQIPS KYVMDQEFSR RLSMSEVKDD NSATKTLSAA ARRSFFRRKH KHKRSGSKDG KDLLALDAFS SDSIPLFEDS VSLAYQRVQK VDCTALRPVL ILGPLLDVVK EMLVNEAPGK FCRCPLEVMK ASQQAIERGV KDCLFVDYKR RSGHFDVTTV ASIKEITEKN RHCLLDIAPH AIERLHHMHI YPIVIFIHYK SAKHIKEQRD PIYLRDKVTQ RHSKEQFEAA QKLEQEYSRY FTGRCAWAWS GEQAAGKEAE AS (1569-1919 aa encoded by B C002915)
May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: discs large homolog 5; discs large protein LP-DLG; Discs large protein P-dlg; discs, large homolog 5; Disks large homolog 5; large type of P-DLG; Placenta and prostate DLG
Gene Aliases: 4933429D20Rik; DLG5; KIAA0583; LP-DLG; mKIAA0583; P-DLG5; PDLG; T25557
UniProt ID: (Human) Q8TDM6, (Mouse) E9Q9R9
Entrez Gene ID: (Human) 9231, (Mouse) 71228
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support