Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LDIAPHAIER LHHMHIYPIV IFIHYKSAKH IKEQRDPIYL RDKVTQRHSK EQFEAAQKLE QEYSRYFTGV IQGGALSSIC TQILAMVNQE QNKVLWIPAC PL
May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: discs large homolog 5; discs large protein LP-DLG; Discs large protein P-dlg; discs, large homolog 5; Disks large homolog 5; large type of P-DLG; Placenta and prostate DLG
Gene Aliases: DLG5; KIAA0583; LP-DLG; P-DLG5; PDLG
UniProt ID: (Human) Q8TDM6
Entrez Gene ID: (Human) 9231
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support