Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MKGLSGSRSH HHGVTCDSAC DSLSHHSDRK PYLLSPVEHH PADHPYYTQR NSFQAECVGP FSDPLASSTF PRRHYTSQQE LKDECALVPR TLATKANRIP
Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAP-1; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; Disks large-associated protein 1; Guanylate kinase-associated protein; hGKAP; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; SAP90/PSD-95-associated protein 1; SAPAP1
Gene Aliases: DAP-1; DAP-1-ALPHA; DAP-1-BETA; DAP1; DLGAP1; DLGAP1A; DLGAP1B; GKAP; SAPAP1
UniProt ID: (Human) O14490
Entrez Gene ID: (Human) 9229
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support