Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSGCRCLHL VCLLCILGAP GQPVRADDCS SHCDLAHGCC APDGSCRCDP GWEGLHCERC VRMPGCQHGT CHQPWQCICH SGWAGKFCDK DEHICTTQSP CQNGGQCMYD GGGEYHCVCL PGFHGRDCER KAGPCEQAGS PCRNGGQCQD DQGFALNFTC RCLVGFVGAR CEVNVDDCLM RPCANGATCL DGINRFSCLC PEGFAGRFCT INLDDCASRP CQRGARCRDR VHDFDCLCPS GYGGKTCELV LPVPDPPTTV DTPLGPTSAV VVPATGPAPH SAGAGLLRIS VKEVVRRQEA GLGEPSLVAL VVFGALTAAL VLATVLLTLR AWRRGVCPPG PCCYPAPHYA PACQDQECQV SMLPAGLPLP RDLPPEPGKT TAL
Regulates adipogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: delta-like 2 homolog; DLK-2; EGF-like protein 9; EGF-like-domain, multiple 9; Epidermal growth factor-like protein 9; Protein delta homolog 2
Gene Aliases: DLK-2; DLK2; EGFL9; UNQ2903/PRO28633
UniProt ID: (Human) Q6UY11
Entrez Gene ID: (Human) 65989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support