Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The HMD1-5 monoclonal antibody reacts with mouse Delta-like 1, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g. Notch 1-4), Delta-like 1 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Delta-like 1 is expressed by thymic stromal cells, as well as on macrophages, dendritic cells, and stromal cells in the spleen. This protein is also expressed in nonhematopoietic tissues such as the lung and brain. Delta-like 1 has been linked to B cell development and differentiation, especially marginal zone and plasma cells, regulation of splenic dendritic cells, and leukemogenesis.
Applications Reported: This HMD1-5 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This HMD1-5 antibody has been tested by flow cytometric analysis on mouse splenocytes and transfected cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: delta like-1; Delta-like protein 1; Delta1; Drosophila Delta homolog 1; OTTHUMP00000017690
Gene Aliases: Delta1; Dll1
UniProt ID: (Mouse) Q61483
Entrez Gene ID: (Mouse) 13388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support