Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Hamster anti Mouse Delta-Like Protein 4 antibody, clone HMD4-2 recognizes mouse Delta-like protein 4 (DLL4), one of the five major ligands of the Notch signalling pathway, which is activated through the binding of specific ligands to the Notch receptors Notch 1-4.
Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delta-like protein 4; Delta4; Drosophila Delta homolog 4; MGC126344
Gene Aliases: Delta4; Dll4
UniProt ID: (Mouse) Q9JI71
Entrez Gene ID: (Mouse) 54485
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support