Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ARESYGFNN IVGYPKEERF AGRPTQRVSG LYQRLESKCS MGFHAGWEQP HWFYKPGQDT QYRPSFRRTN WFEPVGSEYK QVMQRVAVTD LSPFGKFNIK GQDSIRLLDH LFANVIPKVG FTNISHMLTP KGRVYAELTV SHQSPGEFLL ITGSGSELHD LRWIEEEAVK GGYDVEIKNI TDELGVLGVA GPQARKVLQK LTSEDLSDDV FKFLQTKSLK VSNIPVTAIR ISYTGELGWE LYHRREDSVA LYDAIMNAGQ EEGIDNFGTY AMNALRLEKA FRAWGLEMNC DTNPLEAGLE YFVKLNKDQN SCFAHFKEEN GWVSRWAIRP Y (284-613 aa encoded by BC022388)
This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethylglycine dehydrogenase, mitochondrial; EC 1.5.99.2; M2GD; ME2GLYDH
Gene Aliases: 1200014D15Rik; AI787269; DMGDH; DMGDHD; ME2GLYDH
UniProt ID: (Human) Q9UI17, (Mouse) Q9DBT9
Entrez Gene ID: (Human) 29958, (Mouse) 74129, (Rat) 245961
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support