Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: DEGVPEEARD FIQRLLCPPE TRLGRGGAGD FRTHPFFFGL DWDGLRDSVP PFTPDFEGAT DTCNFDLVED GLTAMVSGGG ETLSDIREGA PLGVHLPFVG YSYSCMALRD SEVPGPTP
DMPK is a serine/threonine kinase expressed in various muscle and nerve tissues. Mutations in the untranslated region of the DMPK gene are a characteristic of myotonic dystrophy. The protein encoded by this gene is a serine-threonine kinase that is closely related to other kinases that interact with members of the Rho family of small GTPases. Substrates for this enzyme include myogenin, the beta-subunit of the L-type calcium channels, and phospholemman. The 3' untranslated region of this gene contains 5-37 copies of a CTG trinucleotide repeat. Expansion of this unstable motif to 50-5,000 copies causes myotonic dystrophy type I, which increases in severity with increasing repeat element copy number. Repeat expansion is associated with condensation of local chromatin structure that disrupts the expression of genes in this region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DM protein kinase; DM-kinase; DM1 protein kinase; DMK; DMPK; dystrophia myotonica-protein kinase; MT-PK; myotonic dystrophy associated protein kinase; Myotonic dystrophy protein kinase; myotonin protein kinase A; Myotonin-protein kinase; thymopoietin homolog
Gene Aliases: DM; DM1; DM1PK; DMK; DMPK; MDPK; MT-PK
UniProt ID: (Human) Q09013
Entrez Gene ID: (Human) 1760
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support