Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MPNDEAFSKP STPSEAPHAP GVPPQGRAGG FGKASGALVG AASGSSAGGS SRGGGSGSGA SDLGAGSKKS PRLPKCARCR NHGYASPLKG HKRFCMWRDC QCKKCNLIAE RQRVMAAQVA LRRQQAQEEE LGISHPIPLP SAAELLVKRE NNGSNPCLMT ECSGTSQPPP ASVPTTAASE GRMVIQDIPA VTSRGHVENT PDLVSDSTYY SSFYQPSLFP YYNNLYNCPQ YSMALAADSA SGEVGNPLGG SPVKNSLRGL PGPYVPGQTG NQWQMKNMEN RHAMSSQYRM HSYYPPPSYL GQSVPQFFTF EDAPSYPEAR ASVFSPPSSQ DSGLVSLSSS SPISNKSTKA VLECEPASEP SSFTVTPVIE EDE
This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DM domain expressed in testis 1; DM domain expressed in testis protein 1; Doublesex- and mab-3-related transcription factor 1
Gene Aliases: CT154; DMRT1; DMT1
UniProt ID: (Human) Q9Y5R6
Entrez Gene ID: (Human) 1761
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support