Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KQEESPSDLA SAYVTEGLES PTIEEQVDQT IDDETILIVP SPHGFIQASD VIDTESVLPL TTLTDPILQH HQEESNIIGS SLGSPVSEDS KDVEDLVNCH
This gene encodes a transcription factor that contains a cyclin D-binding domain, three central Myb-like repeats, and two flanking acidic transactivation domains at the N- and C-termini. The encoded protein is induced by the oncogenic Ras signaling pathway and functions as a tumor suppressor by activating the transcription of ARF and thus the ARF-p53 pathway to arrest cell growth or induce apoptosis. It also activates the transcription of aminopeptidase N and may play a role in hematopoietic cell differentiation. The transcriptional activity of this protein is regulated by binding of D-cyclins. This gene is hemizygously deleted in approximately 40% of human non-small-cell lung cancer and is a potential prognostic and gene-therapy target for non-small-cell lung cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cyclin D binding myb-like transcription factor 1; cyclin D-binding Myb-like protein; Cyclin-D-binding Myb-like transcription factor 1; Cyclin-D-interacting Myb-like protein 1; FLJ41265; hDMP1; hDMTF1
Gene Aliases: DMP1; DMTF; DMTF1; hDMP1; MRUL
UniProt ID: (Human) Q9Y222
Entrez Gene ID: (Human) 9988
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support