Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a protein with 12 WD domains. Proteins with WD domains are involved in many functions including participation in signal transduction pathways. Participation of the encoded protein in regulation of the Notch signaling pathway has been demonstrated in vitro using several human cell lines. A gene encoding a similar protein is located on chromosome 5. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dmx-like 2; Dmx-like 2, pseudogene 1; DmX-like protein 2; rabconnectin 3; Rabconnectin-3
Gene Aliases: 6330586A12; 6430411K14Rik; DMXL2; Dmxl2-ps1; E130119P06Rik; KIAA0856; mKIAA0856; PEPNS; RC3
UniProt ID: (Human) Q8TDJ6, (Mouse) Q8BPN8
Entrez Gene ID: (Human) 23312, (Pig) 100155516, (Rat) 315676, (Mouse) 235380
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support