Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; ligase 1; ligase I, DNA, ATP-dependent; MGC117397; MGC130025; Polydeoxyribonucleotide synthase [ATP] 1
Gene Aliases: AL033288; Lig-1; LIG1; LigI
UniProt ID: (Human) P18858, (Mouse) P37913
Entrez Gene ID: (Human) 3978, (Bovine) 100124507, (Mouse) 16881, (Rat) 81513
Molecular Function:
DNA ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support