Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MEIVYVYVKK RSEFGKQCNF SDRQAELNID IMPNPELAEQ FVERNPVDTG IQCSISMSEH EANSERFEME TRGVNHVEGG WPKDVNPLEL EQTIRFRKKV EKDENYVNAI MQLGSIMEHC IKQNNAIDIY EEYFNDEEAM EVMEEDPSAK TINVFRDPQE IKRAATHLSW HPDGNRKLAV AYSCLDFQRA PVGMSSDSYI WDLENPNKPE LALKPSSPLV TLEFNPKDSH VLLGGCYNGQ IACWDTRKGS LVAELSTIES SHRDPVYGTI WLQSKTGTEC FSASTDGQVM WWDIRKMSEP TEVVILDITK KEQLENALGA ISLEFESTLP TKFMVGTEQG IVISCNRKAK TSAEKIVCTF PGHHGPIYAL QRNPFYPKNF LTVGDWTARI WSEDSRESSI MWTKYHMAYL TDAAWSPVRP TVFFTTRMDG TLDIWDFMFE QCDPTLSLKD NGCLIACGSQ LGTTTLLEVS PGLSTLQRNE KNVASSMFER ETRREKILEA RHREMRLKEK GKAEGRDEEQ TDEELAVDLE ALVSKAEEEF FDIIFTELKK KEADAIKLTP VPQQPSPEED QVVEEGEEAA GEEGDEEVEE DLA
The protein encoded by this gene belongs to the dynein intermediate chain family, and is part of the dynein complex of respiratory cilia and sperm flagella. Mutations in this gene are associated with primary ciliary dyskinesia type 9. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal dynein intermediate chain 2; Dynein axonemal intermediate chain 2; dynein, axonemal, intermediate chain 2; dynein, axonemal, intermediate polypeptide 2
Gene Aliases: CILD9; DIC2; DNAI2
UniProt ID: (Human) Q9GZS0
Entrez Gene ID: (Human) 64446
Molecular Function:
microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support