Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVKETTYYD VLGVKPNATQ EELKKAYRKL ALKYHPDKNP NEGEKFKQIS QAYEVLSDAK KRELYDKGGE QAIKEGGAGG GFGSPMDIFD MFFGGGGRMQ RERRGKNVVH QLSVTLEDLY NGATRKLALQ KNVICDKCEG RGGKKGAVEC CPNCRGTGMQ IRIHQIGPGM VQQIQSVCME CQGHGERISP KDRCKSCNGR KIVREKKILE VHIDKGMKDG QKITFHGEGD QEPGLEPGDI IIVLDQKDHA VFTRRGEDLF MCMDIQLVEA LCGFQKPIST LDNRTIVITS HPGQIVKHGD IKCVLNEGMP IYRRPYEKGR LIIEFKVNFP ENGFLSPDKL SLLEKLLPER KEVEETDEMD QVELVDFDPN QERRRHYNGE AYEDDEHHPR GGVQCQTS (1-397 aa encoded by BC008182)
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ homolog subfamily A member 1; DnaJ protein homolog 2; h HSJ-2; hDj-2; Heat shock 40 kDa protein 4; Heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ-2; Human DnaJ protein 2; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7
Gene Aliases: DJ-2; DjA1; DNAJ2; DNAJA1; hDJ-2; HDJ2; HSDJ; HSJ-2; HSJ2; HSPF4; NEDD7
UniProt ID: (Human) P31689, (Mouse) P63037
Entrez Gene ID: (Human) 3301, (Mouse) 15502
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support