Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MIPLLLAAL LCVPAGALTC YGDSGQPVDW FVVYKLPALR GSGEAAQRGL QYKYLDESSG GWRDGRALIN SPEGAVGRSL QPLYRSNTSQ LAFLLYNDQP PQPSKAQDSS MRGHTKGVLL LDHDGGFWLV HSVPNFPPPA SSAAYSWPHS ACTYGQTLLC VSFPFAQFSK MGKQLTYTYP WVYNYQLEGI FAQEFPDLEN VVKGHHVSQE PWNSSITLTS QAGAVFQSFA KFSKFGDDLY SGWLAAALGT NLQVQFWHKT VGILPSNCSD IWQVLNVNQI AFPGPAGPSF NSTEDHSKWC VSPKGPWTCV GDMNRNQGEE QRGGGTLCAQ LPALWKAFQP LVKNYQPCNG MARKPSRAYK I (1-360 aa encoded by BC010419)
DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codominant alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid DNase; deoxyribonuclease; Deoxyribonuclease II alpha; Deoxyribonuclease-2-alpha; DNase II alpha; DNase II, lysosomal; Lysosomal DNase II; R31240_2
Gene Aliases: DNASE2; DNASE2A; DNL; DNL2
UniProt ID: (Human) O00115, (Rat) Q9QZK8
Entrez Gene ID: (Human) 1777, (Rat) 171575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support