Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
2 µg/mL of MA5-45296 was sufficient for detection of Dnmt1 in 10 µg of mouse ES cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 180kDa. It will cross-react with mouse DNMT1.
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CXXC-type zinc finger protein 9; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase HsaI; DNA methyltransferase MmuI; DNA MTase HsaI; DNA MTase MmuI; Dnmt1; FLJ16293; MCMT; MGC104992
Gene Aliases: ADCADN; AIM; cb1075; cb983; CXXC9; ddn; dmnt1; DNMT; DNMT1; Dnmt1o; fb05h01; HSN1E; m.HsaI; m.MmuI; MCMT; Met-1; Met1; MommeD2; mta; MTase; Uim; wu:fb05h01
UniProt ID: (Human) P26358, (Mouse) P13864
Entrez Gene ID: (Human) 1786, (Mouse) 13433, (Zebrafish) 30430
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support