Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: SLKIAAFNIQ TFGETKMSNA TLVSYIVQIL SRYDIALVQE VRDSHLTAVG KLLDNLNQDA PDTYHYVVSE PLGRNSYKER YLFVYRPDQV SAVDSYYYDD GCEPCGNDTF NREPAIVRFF SRFTEVREFA IVPLHAAPGD AVAEIDALYD VYLDVQEKWG LEDVMLMGDF NAGCSYVRPS QWSSIRLWTS PTFQWLIPDS ADTTATPTHC AYDRIVVAGM LLRGAVVPDS ALPFNFQAAY GLSDQLAQAI SDHYPVEVML K
Internucleosomal DNA fragmentation following the activation of endonucleases is the common end point of apoptosis. DNase I, a Ca2+/Mg2+-dependent endonuclease ubiquitously expressed in mammalian tissues, has been implicated to mediate internucleosomal DNA degradation in human cells undergoing apoptosis. DNase I is highly polymorphic, and at least six alleles of DNase I are known. DNase II, the ubiquitously expressed acidic deoxyribonuclease, acts downstream of caspase activation and may also induce DNA digestion during apoptosis. DNase I cleaves DNA to 5'-phosphodinucleotide and 5'-phospho- oligonucleotide end-products, whereas DNase II cleaves DNA to 3'-phosphomononucleotide and 3'-phosphooligonucleotide end-products. The mechanism by which DNase II cuts DNA is similar to DNase I, which produces nicks rather than double-strand cuts. DNase II is usually present in cytoplasm of epithelial cells, but it appears concentrated in the nuclei of lens fibers. In contrast, DNase I is always concentrated in nuclei of epithelial and fiber cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deoxyribonuclease; Deoxyribonuclease 1; Deoxyribonuclease I; Deoxyribonuclease-1; Deoxyribonuclease1; DeoxyribonucleaseI; DNASE 1; DNase I; DNase I lysosomal; DNase I, lysosomal; DNL 1; Dornase alfa; FLJ38093; human urine deoxyribonuclease I
Gene Aliases: AI788650; DNASE1; DNaseI; DNL1; DRNI
UniProt ID: (Human) P24855, (Mouse) P49183, (Rat) P21704
Entrez Gene ID: (Human) 1773, (Mouse) 13419, (Rat) 25633
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support