Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MCVCAEDGKT LPVAVPIEDM QRIHLRFMFR HRSSLESKDK GEKNFAMSYV KLMKEDGTTL HDGFHDLVVL KGDSKKMEDA SAYLTLPSYR HHVENKGATL SRSSSSVGGL SVSSRDVFSI STLVCSTKLT QNVGLLGLLK WRMKPQLEES MRRLFESINN LMKSQYKTTI LLQVAALKYI PSVLHDVEMV FDAKLLSQLL YEFYTCIPPV KLQKQKVQSM NEIVQSNLFK KQECRDILLP VITKELKELL EQKDDMQHQV LERKYCVELL NSILEVLSYQ DAAFTYHHIQ EIMVQLLRTV NRTVITMGRD HILIGACCNC GFPSSTPHSL IQWLWG
The DOCK2 gene encodes a hematopoietic cell-specific CDM family protein that is indispensable for lymphocyte chemotaxis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dedicator of cyto-kinesis 2; Dedicator of cytokinesis protein 2; FLJ46592
Gene Aliases: DOCK2; IMD40; KIAA0209
UniProt ID: (Human) Q92608
Entrez Gene ID: (Human) 1794
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support