Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SRC2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dedicator of cytokinesis 4; Dedicator of cytokinesis protein 4
Gene Aliases: 5330406C03; 6330411N01Rik; AF263288; C030023J22; DOCK4; KIAA0716; mKIAA0716; RGD1561724
UniProt ID: (Human) Q8N1I0, (Mouse) P59764
Entrez Gene ID: (Human) 9732, (Mouse) 238130, (Rat) 366608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support