Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1200017A24Rik; Dedicator of cytokinesis protein 8; epididymis luminal protein 205
Gene Aliases: DOCK8; HEL-205; MRD2; ZIR8
UniProt ID: (Human) Q8NF50
Entrez Gene ID: (Human) 81704
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support