Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
DOCK9 is a guanine nucleotide-exchange factor (GEF) that activates CDC42 by exchanging bound GDP for free GTP. Overexpression of DOCK9 induces filopodia formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cdc42 guanine nucleotide exchange factor zizimin-1; Dedicator of cytokinesis protein 9; Zizimin-1
Gene Aliases: DOCK9; KIAA1058; ZIZ1; ZIZIMIN1
UniProt ID: (Human) Q9BZ29
Entrez Gene ID: (Human) 23348
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support