Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive control: Human gastrointestinal stromal tumor tissue, human gallbladder tissue, human breast tissue.
Predicted band size: 114 kDa
Subcellular Location: Cytoplasm, Cell membrane.
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anoctamin 1, calcium activated chloride channel; Anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; Discovered on gastrointestinal stromal tumors protein 1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); Tumor-amplified and overexpressed sequence 2
Gene Aliases: ANO1; DOG1; ORAOV2; TAOS2; TMEM16A
UniProt ID: (Human) Q5XXA6
Entrez Gene ID: (Human) 55107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support