Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MVTEQEVDAI GQTLVDPKQP LQARFRALFT LRGLGGPGAI AWISQAFDDD SALLKHELAY CLGQMQDARA IPMLVDVLQD TRQEPMVRHE AGEALGAIGD PEVLEILKQY SSDPVIEVAE TCQLAVRRLE WLQQHGGEPA AGPYLSVDPA PPAEERDVGR LREALLDESR PLFERYRAMF ALRNAGGEEA ALALAEGLHC GSALFRHEVG YVLGQLQHEA AVPQLAAALA RCTENPMVRH ECAEALGAIA RPACLAALQA HADDPERVVR ESCEVALDMY EHETGRAFQY ADGLEQLRGA PS
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deoxyhypusine dioxygenase; deoxyhypusine dioxygenase {ECO:0000255|HAMAP-Rule:MF_03101}; Deoxyhypusine hydroxylase; deoxyhypusine hydroxylase {ECO:0000255|HAMAP-Rule:MF_03101}; Deoxyhypusine monooxygenase; deoxyhypusine monooxygenase {ECO:0000255|HAMAP-Rule:MF_03101}; DOHH; DOHH {ECO:0000255|HAMAP-Rule:MF_03101}; hDOHH; HEAT-like (PBS lyase) repeat containing 1; HEAT-like repeat-containing protein 1
Gene Aliases: 1110033C18Rik; AI585913; AL022869; DOHH; hDOHH; HLRC1; RGD1304783
UniProt ID: (Human) Q9BU89, (Rat) Q5PPJ4, (Mouse) Q99LN9
Entrez Gene ID: (Human) 83475, (Rat) 314644, (Mouse) 102115
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support