Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: IGTVGIASYD YHQGSGTFLF QAGSGIYHVK DGGPQGFTQQ PLRPNLVETS CPNIRMDPKL CPADPDWIAF IHSNDIWISN IVTR
This gene encodes a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The encoded protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dipeptidyl peptidase 8; Dipeptidyl peptidase IV-related protein 1; dipeptidyl peptidase IV-related protein-1; Dipeptidyl peptidase VIII; dipeptidyl-peptidase 8; DP8; DPP VIII; DPRP-1; Prolyl dipeptidase DPP8
Gene Aliases: DP8; DPP8; DPRP-1; DPRP1; MST097; MSTP097; MSTP135; MSTP141
UniProt ID: (Human) Q6V1X1
Entrez Gene ID: (Human) 54878
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support