Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAPVLSKDSA DIESILALNP RTQTHATLCS TSAKKLDKKH WKRNPDKNCF NCEKLENNFD DIKHTTLGER GALREAMRCL KCADAPCQKS CPTNLDIKSF ITSIANKNYY
Involved in pyrimidine base degradation. Catalyzes the reduction of uracil and thymine. Also involved the degradation of the chemotherapeutic drug 5-fluorouracil.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHPDHase; Dihydropyrimidine dehydrogenase [NADP(+)]; Dihydrothymine dehydrogenase; Dihydrouracil dehydrogenase
Gene Aliases: DHP; DHPDHASE; DPD; DPYD
UniProt ID: (Human) Q12882
Entrez Gene ID: (Human) 1806
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support