Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IVVCSIRKS SRTLKKGPRQ DPSAIVEKAG LKKSMTPTQN REKWIYYCNG HGIDILKLVA AQVGSQWKDI YQFLCNASER EVAAFSNGYT ADHERAYAAL QHWTIRGPEA SLAQLISALR QHRRNDVVEK IRGLMEDTTQ LETDKLALPM SPSPLSPSPI PSPNAKLENS ALLTVEPSPQ DKNKGFFVDE SEPLLRCDST SSGSSALSRN GSFITKEKKD TVLRQVRLDP CDLQPIFDDM LHFLNPEELR VIEEIPQAED KLDRLFEIIG VKSQEASQTL LDSVYSHLPD LL (365-655 aa encoded by BC021572)
DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kappa-B and JNK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD358; Death receptor 6; Death receptor-6; MGC31965; sCD358; sDR6; soluble CD358; soluble DR6; TNFR-related death receptor 6; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21
Gene Aliases: BM-018; CD358; DR6; TNFRSF21; UNQ437/PRO868
UniProt ID: (Human) O75509
Entrez Gene ID: (Human) 27242
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support