Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: HeLa, C6, SH-SY5Y, U-251MG, Mouse brain
Immunogen sequence: MASLSQLSGH LNYTCGAENS TGASQARPHA YYALSYCALI LAIVFGNGLV CMAVLKERAL QTTTNYLVVS LAVADLLVAT LVMPWVVYLE VTGGVWNFSR
This gene encodes the D3 subtype of the five (D1-D5) dopamine receptors. The activity of the D3 subtype receptor is mediated by G proteins which inhibit adenylyl cyclase. This receptor is localized to the limbic areas of the brain, which are associated with cognitive, emotional, and endocrine functions. Genetic variation in this gene may be associated with susceptibility to hereditary essential tremor 1. Alternative splicing of this gene results in transcript variants encoding different isoforms, although some variants may be subject to nonsense-mediated decay (NMD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D(3) dopamine receptor; D3 receptor; D3nf; Dopamine 3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopaminergic receptor D3; essential tremor 1
Gene Aliases: D3DR; D3R; DRD3; ETM1; FET1
UniProt ID: (Human) P35462, (Rat) P19020, (Mouse) P30728
Entrez Gene ID: (Human) 1814, (Rat) 29238, (Mouse) 13490
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support