Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Isoform 1 may function as a nuclear receptor coactivator, enhancing transcription through other coactivators such as NCOA6 and CITED1. Isoform 2, functions as a transcriptional repressor, modulating transcriptional activities of coactivators including isoform 1, NCOA6 and CITED1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: down regulated in metastasis; Down-regulated in metastasis protein; NNP73; Novel nucleolar protein 73; Protein Key-1A6; Small subunit processome component 20 homolog; UTP20 small subunit (SSU) processome component; UTP20 small subunit processome component; UTP20, small subunit (SSU) processome component, homolog
Gene Aliases: 1A6/DRIM; 3830408P06Rik; AA617408; DRIM; mDRIM; mDRIN; UTP20
UniProt ID: (Human) O75691
Entrez Gene ID: (Human) 27340, (Mouse) 70683
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support