Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CGGARICYIF HETFGRTLES VDPLGGLNTI DILTAIRNAT GPRPALFVPE VSFELLVKRQ IKRLEEPSLR CVELVHEEMQ RIIQHCSNYS TQELLRFPKL HDAIVEVVTC LLRKRLPVTN EMVHNLVAIE LAYINTKHPD FADACGLMNN NIEEQRRNRL ARELPSAVSR DKLIQDSRRE TKNVASGGGG VGDGVQEPTT GNWRGMLKTS KAEELLAEEK SKPIPIMPAS PQKGHAVNLL DVPVPVARKL SAREQRDCEV IERLIKSYFL IVRKNIQDSV PKAVMHFLVN HVKDTLQSEL VGQLYKSSLL DDLLTESEDM AQRRKEAADM LKALQGASQI IAEIRETHLW
A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-terminal region; Dnm1p/Vps1p-like protein; DVLP; Dymple; dynamin 1-like; Dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-like protein; dynamin-like protein 1; Dynamin-like protein 4; Dynamin-like protein IV; Dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; FLJ41912; HdynIV; N-terminal region; VPS1
Gene Aliases: 6330417M19Rik; AI450666; DLP1; DNM1L; Dnml1; Dnmlp1; DRP1; DVLP; DYMPLE; EMPF; HDYNIV; python
UniProt ID: (Human) O00429, (Rat) O35303, (Mouse) Q8K1M6
Entrez Gene ID: (Human) 10059, (Rat) 114114, (Mouse) 74006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support