Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: CKPKMGYTDI LAVDPDEPVH GAPFYFSLPN TSPEISRLWS LTKVNDTAAR LSYQKNAGFQ EYTIPITVKD RAGQAATKLL RVNLCECTHP TQCRATSRST
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARVC10; ARVD10; Cadherin family member 3; CDHF5; CDHF6; CMD1BB; Desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; HT-CP
Gene Aliases: CDHF3; DSC; DSC1; DSC2; DSC3; DSC4; HT-CP
UniProt ID: (Human) Q14574
Entrez Gene ID: (Human) 1825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support