Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: LGEESPLETA ASFFHNVDVP FEETVVDGVH GAFIRQRDGL YKMYWMDDTG YSEKATFASV TYPRGYPYNG TNYVNVTMHL RSPITRAAYL FIGPSIDVQS
Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitin-glucuronate 5-epimerase; Dermatan-sulfate epimerase; DS epimerase; SART-2; Squamous cell carcinoma antigen recognized by T-cells 2
Gene Aliases: DS-epi1; DSE; DSEP; DSEPI; EDSMC2; SART-2; SART2
UniProt ID: (Human) Q9UL01
Entrez Gene ID: (Human) 29940
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support