Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine
Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (approximately 450 kDa) consisting of 24 subunits of two types, H (heavy) and L (light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA379J5.3; bA555E18.1; D-aminoacyl-tRNA deacylase 1; D-tyrosyl-tRNA deacylase 1 homolog; DNA-unwinding element-binding protein B; DTD; DUE-B; Gly-tRNA(Ala) deacylase; Histidyl-tRNA synthase-related; histidyl-tRNA synthetase 2; MGC119131; MGC41905
Gene Aliases: C20orf88; DTD1; DUE-B; DUEB; HARS2; pqn-68
UniProt ID: (Human) Q8TEA8
Entrez Gene ID: (Human) 92675
Molecular Function:
esterase
hydrolase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support