Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SHLRPPSPLL VRVYKSGPRV RRKLESYFQS SKSSGGGECT VSTQEHEAPG TFRVEFSERA AKERVLKKGE HQILVDEKPV PIFLVPTENS IKKNTRPQIS SLTQSQAE
Component of the shelterin complex (telosome) that is involved in the regulation of telomere length and protection. Shelterin associates with arrays of double-stranded TTAGGG repeats added by telomerase and protects chromosome ends. Without its protective activity, telomeres are no longer hidden from the DNA damage surveillance and chromosome ends are inappropriately processed by DNA repair pathways. Promotes binding of POT1 to single-stranded telomeric DNA. Modulates the inhibitory effects of POT1 on telomere elongation. The ACD-POT1 heterodimer enhances telomere elongation by recruiting telomerase to telomeres and increasing its processivity. May play a role in organogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-lymphoma- and BAL-associated protein; deltex 3 like, E3 ubiquitin ligase; deltex 3-like; E3 ubiquitin-protein ligase DTX3L; Protein deltex-3-like; rhysin 2; Rhysin-2; Rhysin2; RING-type E3 ubiquitin transferase DTX3L
Gene Aliases: BBAP; DTX3L
UniProt ID: (Human) Q8TDB6
Entrez Gene ID: (Human) 151636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support