Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPKLQGFEFW SRTLRGARHV VAPMVDQSEL AWRLLSRRHG AQLCYTPMLH AQVFVRDANY RKENLYCEVC PEDRPLIVQF CANDPEVFVQ AALLAQDYCD AIDLNLGCPQ MIAKRGHYGA FLQDEWDLLQ RMILLAHEKL SVPVTCKIRV FPEIDKTVRY AQMLEKAGCQ LLTVHGRTKE QKGPLSGAAS WEHIKAVRKA VAIPVFANGN IQCLQDVERC LRDTGVQGVM SAEGNLHNPA LFEGRSPAVW ELAEEYLDIV REHPCPLSYV RAHLFKLWHH TLQVHQELRE ELAKVKTLEG IAAVSQELKL RCQEEISRQE GAKPTGDLPF HWICQPYIRP GPREGSKEKA GARSKRALEE EEGGTEVLSK NKQKKQLRNP HKTFDPSLKP KYAKCDQCGN PKGNRCVFSL CRGCCKKRAS KETADCPGHG LLFKTKLEKS LAWKEAQPEL QEPQPAAPGT PGGFSEVMGS ALA
Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs. Target protein has a region of similarity to tRNA dihydrouridine synthase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dihydrouridine synthase 1-like; tRNA-dihydrouridine synthase 1-like; tRNA-dihydrouridine(16/17) synthase [NAD(P)(+)]-like
Gene Aliases: DUS1; DUS1L; PP3111
UniProt ID: (Human) Q6P1R4
Entrez Gene ID: (Human) 64118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support