Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MPPSPLDDRV VVALSRPVRP QDLNLCLDSS YLGSANPGSN SHPPVIATTV VSLKAANLTY MPSSSGSARS LNCGCSSASC CTVATYDKDN QAQTQAIAAG TTTTAIGTST TCPANQMVNN NENTGSLSPS SGVGSPVSGT PKQLASIKII YPNDLAKKMT KCSKSHLPSQ GPVIIDCRPF MEYNKSHIQG AVHINCADKI SRRRLQQGKI TVLDLISCRE GKDSFKRIFS KEIIVYDENT NEPSRVMPSQ PLHIVLESLK REGKEPLVLK GGLSSFKQNH ENLCDNSLQL QECREVGGGA
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity phosphatase MKP-5; Dual specificity protein phosphatase 10; MAP kinase phosphatase 5; Mitogen-activated protein kinase phosphatase 5; serine/threonine specific protein phosphatase
Gene Aliases: 2610306G15Rik; AI158871; DUSP10; MKP-5; MKP5
UniProt ID: (Human) Q9Y6W6, (Mouse) Q9ESS0
Entrez Gene ID: (Human) 11221, (Mouse) 63953, (Rat) 63995
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support