Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALLTALDDT LPEEAQGPGR RMILLSTPSQ SDALRACFER NLYPGIATKE QLAQGIDIPE PRVQIWFQNE RSCQLRQHRR QSRPWPGRRD PQKGRRKRTA ITGSQTALLL RAFEKDRFPG IPAREELARE TGLPESRIQL WFQNRRARHW GQSGRAPTQA SIRCNAAPIG
The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX3, resides within these 3.3-kb repeated elements (Beckers et al., 2001 [PubMed 11245978]). See DUX4 (MIM606009).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: double homeobox, 3
Entrez Gene ID: (Human) 26582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support