Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LGQGYPYQY PGPPPCFPPA YQDPGFSYGS GSTGSQQSEG SKSSGSTRSS RRAPGREKER RAAGAGGSGS ESDHTAPSGV GSSWRERPAG QLSRGSSPRS QASATAPGLP PPHPTTKAYT VVGGPPGGPP VRELA (283-416 aa encoded by BC050454)
DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dishevelled 1 (homologous to Drosophila dsh); dishevelled, dsh homolog 1; Dishevelled-1; Dishevelled-1-like; DSH homolog 1; DSH homolog 1-like; MGC54245; RP5-890O3.5; Segment polarity protein dishevelled homolog DVL-1
Gene Aliases: DRS2; DVL; DVL1; DVL1L1; DVL1P1; mKIAA4029
UniProt ID: (Human) O14640, (Mouse) P51141
Entrez Gene ID: (Human) 1855, (Mouse) 13542
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support