Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA36D19.5; BC019250 protein; DPY30 domain containing 1; DPY30 domain-containing protein 1; FLJ43920; OTTHUMP00000019963
Gene Aliases: 1700029M23Rik; DPY30D1; DYDC1; RSD-9; RSD9
UniProt ID: (Human) Q8WWB3, (Mouse) Q9D9T0
Entrez Gene ID: (Human) 143241, (Mouse) 69496, (Rat) 688433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support