Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total DYNC2LI1.
DYNC2LI1 (dynein, cytoplasmic 2, light intermediate chain 1), also known as LIC3, D2LIC or CGI-60, is a 351 amino acid cytoplasmic protein belonging to the dynein light intermediate chain family. DYNC2LI1 may function as a motor for intraflagellar retrograde transport and in cilia biogenesis. The cytoplasmic dynein complex 2 may be composed of a DYNC2H1 homodimer and a number of DYNC2LI1 light intermediate chains. DYNC2LI1 exists as five alternatively spliced isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 2 light intermediate chain 1; Dynein 2 light intermediate chain; dynein, cytoplasmic 2, light intermediate chain 1; mD2LIC
Gene Aliases: 4933404O11Rik; CGI-60; D2LIC; DYNC2LI1; LIC3; mD2LIC; RGD1310286
UniProt ID: (Human) Q8TCX1, (Mouse) Q8K0T2
Entrez Gene ID: (Human) 51626, (Mouse) 213575, (Rat) 298767
Molecular Function: cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support