Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSDRKAVIKN ADMSEDMQQD AVDCATQAME KYNIEKDIAA YIKKEFDKKY NPTWHCIVGR NFGSYVTHET KH
The directed movement of membrane-bounded organelles is an important process for various cellular functions including membrane transport, secretion, and axonal transport. Cytoplasmic dynein, a ubiquitous minus end-directed microtubule-based motor protein, is believed to be responsible for the retrograde transport of membranous organelles from the synapse to the cell body in fast axonal transport. The cytoplasmic dynein complex is composed of two ~530 kDa heavy chains, and intermediate chains of ~74 kDa and ~53-59 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 8 kDa dynein light chain b; DLC8b; Dynein light chain 2, cytoplasmic; Dynein light chain LC8-type 2; dynein, light chain, LC8-type 2; radial spoke 22 homolog
Gene Aliases: DLC2; DNCL1B; DYNLL2; RSPH22
UniProt ID: (Human) Q96FJ2
Entrez Gene ID: (Human) 140735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support