Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
E3 Ubiquitin ligase proteins mediate ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Able to specifically bind RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAZ interacting protein 3, zinc finger; DAZ interacting zinc finger protein 3; DAZ-interacting protein 3; DAZ-interacting protein 3 homolog; E3 ubiquitin-protein ligase DZIP3; hRUL138; human RNA-binding ubiquitin ligase of 138 kDa; protein phosphatase 1, regulatory subunit 66; RING-type E3 ubiquitin transferase DZIP3; RNA-binding RING-H2 protein-ubiquitin ligase; RNA-binding ubiquitin ligase of 138 kDa; UURF2 ubiquitin ligase; zinc finger DAZ interacting protein 3
Gene Aliases: 2310047C04Rik; 2A-HUB; 6430549P11Rik; A230104G20; DZIP3; hRUL138; KIAA0675; PPP1R66; RGD1563278; UURF2
UniProt ID: (Human) B3KN01, (Mouse) Q7TPV2
Entrez Gene ID: (Human) 9666, (Mouse) 224170, (Rat) 303963
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support