Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAF-1 related transcriptional regulator (NUDR); DEAF1 transcription factor; Deformed epidermal autoregulatory factor 1 homolog; Nuclear DEAF-1-related transcriptional regulator; NUDR; Suppressin; Zinc finger MYND domain-containing protein 5
Gene Aliases: AU042387; C230009B13Rik; DEAF1; MRD24; NUDR; SPN; ZMYND5
UniProt ID: (Human) O75398, (Mouse) Q3UJA4
Entrez Gene ID: (Human) 10522, (Mouse) 54006, (Rat) 83632
Molecular Function:
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support