Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
The antibody detects DELTA-1 from mouse. Activity against Delta-1 from other species has not been determined but is expected based on sequence homology. Expected to cross-react with rat, human and chicken.
Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-Delta-1; delta like-1; delta-like 1; Delta-like protein 1; Delta1; Drosophila Delta homolog 1; H-Delta-1; OTTHUMP00000017690
Gene Aliases: Delta; DELTA1; DL1; DLL1; UNQ146/PRO172
UniProt ID: (Human) O00548, (Mouse) Q61483, (Rat) P97677
Entrez Gene ID: (Human) 28514, (Chicken) 395820, (Mouse) 13388, (Rat) 84010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support