Invitrogen
The envelope protein (E protein) of virus is a large and cysteine-rich protein, which is responsible for a wide range of biological activities, including binding to host cell receptors, fusion and entry into host cells. For instance, the dengue E protein consists of three structurally distinct domains (I, II, and III).
Protein Aliases: Dengue Virus 1; Dengue Virus 2; Dengue Virus 3; Dengue Virus Type 1; Dengue Virus Type 2; Dengue Virus Type 3; DENV; DENV1; DENV2; DENV3