Invitrogen
FIGURE: 1 / 3
Recommended positive controls: Dengue virus 2-infected C6/36, Dengue virus type 2 infected BHK-21, Dengue virus type 3 infected BHK-21.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: Dengue NS4; Dengue NS4 protein