Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DSG1 (Desmoglein 1) is a Protein Coding gene. Diseases associated with DSG1 include Erythroderma, Congenital, With Palmoplantar Keratoderma, Hypotrichosis, And Hyper-Ige and Palmoplantar Keratoderma I, Striate, Focal, Or Diffuse. Among its related pathways are Keratinization and Apoptotic execution phase. Gene Ontology (GO) annotations related to this gene include calcium ion binding and toxic substance binding. An important paralog of this gene is DSG4. DSG2 (Desmoglein 2) is a Protein Coding gene. Diseases associated with DSG2 include Arrhythmogenic Right Ventricular Dysplasia, Familial, 10 and Cardiomyopathy, Dilated, 1Bb. Among its related pathways are Keratinization and Apoptotic execution phase. Gene Ontology (GO) annotations related to this gene include calcium ion binding and cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication. An important paralog of this gene is DSG4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cadherin family member 4; Cadherin family member 5; desmoglein 1; desmoglein 1 beta; desmoglein 1 gamma; desmoglein 2; Desmoglein-1; Desmoglein-2; Desmosomal glycoprotein 1; DG1; DGI; DSG1; DSG1/2; DSG2; HDGC; Pemphigus foliaceus antigen
Gene Aliases: AA408168; CDHF4; CDHF5; D18Ertd293e; DG1; DSG; DSG1; Dsg1b; Dsg1c; DSG2; EPKHE; EPKHIA; HDGC; PPKS1; SPPK1
UniProt ID: (Human) Q02413, (Bovine) Q03763, (Human) Q14126, (Mouse) O55111
Entrez Gene ID: (Human) 1828, (Bovine) 281131, (Human) 1829, (Bovine) 508151, (Rat) 291755, (Mouse) 13511, (Rat) 307562
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support