Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cadherin family member 4; Desmoglein-1; Desmosomal glycoprotein 1; DG1; Pemphigus foliaceus antigen
Gene Aliases: CDHF4; DG1; DSG; DSG1; EPKHE; EPKHIA; PPKS1; SPPK1
UniProt ID: (Human) Q02413
Entrez Gene ID: (Human) 1828
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support