Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: THWHQVRVFH QNVFPNFTVV NVEKPPCFLR KFSPDGRYFI AFSSDQTSLE IYEYQGCQAA EDLLQGYEGE ILSNGNDQRS VNIRGRLFER FFVLLHITNV AANGEHLNRE CS
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
DET1 is a component of the E3 ubiquitin ligase DCX DET1-COP1 complex, which is required for ubiquitination and subsequent degradation of target proteins. The complex is involved in JUN ubiquitination and degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: De-etiolated-1 homolog; DET1 homolog
Gene Aliases: DET1
UniProt ID: (Human) Q7L5Y6
Entrez Gene ID: (Human) 55070
Molecular Function:
ubiquitin-protein ligase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support