Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Small GTPase. Negatively regulates the transcription regulation activity of the APBB1/FE65-APP complex via its interaction with APBB1/FE65.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator of G-protein signaling 1; Dexamethasone-induced Ras-related protein 1; RAS, dexamethasone-induced 1; ras-related protein
Gene Aliases: AGS1; DEXRAS1; MGC:26290; RASD1
UniProt ID: (Human) Q9Y272
Entrez Gene ID: (Human) 51655, (Bovine) 507449
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support