Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: CDSKMKPDR LNYCRHVLAA WALGAKAFYY PEEAGLAFGG PGSSRYLRLE VHYHNPLVIE GRNDSSGIRL YYTAKLRRFN AGIMELGLVY TPVMAIPPRE TAFILTGYCT DKCTQLALPP SGIHIFASQL HTHLTGRKVV TVLVRDGREW EIVNQDNHYS PHFQEIRMLK KVVSVHPGDV LITSCTYNTE DRELATVGGF GILEEMCVNY VHYYPQTQLE LCKSAVDAGF LQKYFHLINR FNNEDVCTCP QASVSQQFTS VPWNSFNRDV LKALYSFAPI SMHCNKSSAV RFQGEWNLQP LPKVISTLEE PTPQCPTSQG RSPAGPTVVS IGGGKG (269-603 aa encoded by BC017174)
The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); Dopamine beta-monooxygenase; Dopamine-Beta-hydroxylase
Gene Aliases: DBH; DBM
UniProt ID: (Human) P09172
Entrez Gene ID: (Human) 1621
Molecular Function:
hydroxylase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support